Product Details
- SNP ID
-
rs11841674
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:99203976 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCAATTATGACCGGGTAAAAGTGC[A/G]TAGGATTAGTGATCATGACTTATTT
- Phenotype
-
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
UBAC2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs140859469] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- UBAC2
- Gene Name
- UBA domain containing 2
- Gene
- UBAC2-AS1
- Gene Name
- UBAC2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details