Product Details
- SNP ID
-
rs4330705
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:143293333 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGGTCTCTCTCTCAGTGCTTCTC[A/C]ACCTTGGCACCGTGGCTGTTTTGGA
- Phenotype
-
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF696
PubMed Links
Gene Details
- Gene
- ZNF696
- Gene Name
- zinc finger protein 696
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_030895.2 |
|
Intron |
|
|
NP_112157.2 |
View Full Product Details