Product Details

SNP ID
rs7183703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:92395112 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTCGCCGGGCCGGGCCCCTCTCC[A/G]GAACCTCCAAAGGGCGGTAGACGAC
Phenotype
MIM: 602546
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ST8SIA2 PubMed Links

Gene Details

Gene
ST8SIA2
Gene Name
ST8 alpha-N-acetyl-neuraminide alpha-2,8-sialyltransferase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006011.3 Intron NP_006002.1
XM_017022642.1 Intron XP_016878131.1
XM_017022643.1 Intron XP_016878132.1

View Full Product Details