Product Details
- SNP ID
-
rs12510437
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:155934079 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACGGCAGCCCCTCCCATCACAGGCT[C/T]GGAGGCCCAGAAGGAAAAAGTGGTT
- Phenotype
-
MIM: 600550
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTSO
PubMed Links
Gene Details
- Gene
- CTSO
- Gene Name
- cathepsin O
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001334.2 |
|
Intron |
|
|
NP_001325.1 |
View Full Product Details