Product Details

SNP ID
rs9654238
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:155343790 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTAATACCAAGAATTTTGCCTACCC[A/G]AGTTAACATTATACAATTACATGTA
Phenotype
MIM: 610070
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP9 PubMed Links

Gene Details

Gene
MAP9
Gene Name
microtubule associated protein 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039580.1 6218 UTR 3 NP_001034669.1
XM_006714306.3 6218 Intron XP_006714369.1
XM_011532253.1 6218 UTR 3 XP_011530555.1
XM_011532254.2 6218 UTR 3 XP_011530556.1
XM_011532255.2 6218 UTR 3 XP_011530557.1
XM_017008616.1 6218 Intron XP_016864105.1
XM_017008617.1 6218 UTR 3 XP_016864106.1
XM_017008618.1 6218 Intron XP_016864107.1

View Full Product Details