Product Details

SNP ID
rs1440798
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:52534696 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAACAGTTTTATTTTGCTTAAAGA[A/C]TCTGTGGACTCCTAATTTACAGTCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM150A PubMed Links

Gene Details

Gene
FAM150A
Gene Name
family with sequence similarity 150 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207413.3 Intron NP_997296.1

View Full Product Details