Product Details

SNP ID
rs12986171
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:10662888 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGCTGTTGCCTGGTCTAAGGGA[A/G]GAGGACCCAGGCATGTAACGGACTG
Phenotype
MIM: 603182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ILF3 PubMed Links

Gene Details

Gene
ILF3
Gene Name
interleukin enhancer binding factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001137673.1 Intron NP_001131145.1
NM_004516.3 Intron NP_004507.2
NM_012218.3 Intron NP_036350.2
NM_017620.2 Intron NP_060090.2
NM_153464.2 Intron NP_703194.1
XM_006722742.2 Intron XP_006722805.1
XM_011527984.2 Intron XP_011526286.2
XM_011527985.2 Intron XP_011526287.1
XM_011527987.1 Intron XP_011526289.1
XM_017026763.1 Intron XP_016882252.1
XM_017026764.1 Intron XP_016882253.1
XM_017026765.1 Intron XP_016882254.1
Gene
ILF3-AS1
Gene Name
ILF3 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details