Product Details
- SNP ID
-
rs11671653
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:10727810 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAACTTCTGCCCGATTTGAATGAAA[A/G]TGCTGAAATTATAGGCCCCTCGGCT
- Phenotype
-
MIM: 602378
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DNM2
PubMed Links
Gene Details
- Gene
- DNM2
- Gene Name
- dynamin 2
- Gene
- MIR638
- Gene Name
- microRNA 638
There are no transcripts associated with this gene.
View Full Product Details