Product Details

SNP ID
rs34626880
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:10729752 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CCCAGCCCGTCCCTTCCTCCAGCTC[C/T]ACCTGGGAGCACCACTGTCACTTCA
Phenotype
MIM: 602378
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNM2 PubMed Links

Gene Details

Gene
DNM2
Gene Name
dynamin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005360.2 Intron NP_001005360.1
NM_001005361.2 Intron NP_001005361.1
NM_001005362.2 Intron NP_001005362.1
NM_001190716.1 Intron NP_001177645.1
NM_004945.3 Intron NP_004936.2

View Full Product Details