Product Details
- SNP ID
-
rs453766
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:11296163 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TGGGCCGCCTTGGACAACCATCAGG[C/G]AGCCAGGACACAGAGGGGCAGAGCA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HSPC102
PubMed Links
Gene Details
- Gene
- HSPC102
- Gene Name
- uncharacterized HSPC102
There are no transcripts associated with this gene.
- Gene
- TSPAN16
- Gene Name
- tetraspanin 16
View Full Product Details