Product Details

SNP ID
rs35082430
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:59584527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATATTACATGCATGAGTGTGTGC[A/C]TGCACAAGTGTGTGCATGAGTGTGT
Phenotype
MIM: 608725
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PHACTR3 PubMed Links

Gene Details

Gene
PHACTR3
Gene Name
phosphatase and actin regulator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199505.1 Intron NP_001186434.1
NM_001199506.1 Intron NP_001186435.1
NM_001281507.1 Intron NP_001268436.1
NM_080672.4 Intron NP_542403.1
NM_183244.1 Intron NP_899067.1
NM_183246.1 Intron NP_899069.1
XM_011528525.2 Intron XP_011526827.1
XM_011528526.2 Intron XP_011526828.1
XM_017027626.1 Intron XP_016883115.1
XM_017027627.1 Intron XP_016883116.1
XM_017027628.1 Intron XP_016883117.1
XM_017027629.1 Intron XP_016883118.1
XM_017027630.1 Intron XP_016883119.1
XM_017027631.1 Intron XP_016883120.1

View Full Product Details