Product Details

SNP ID
rs2341044
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13824648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCACCCTCCTGGCCCCCAGGCCC[A/G]TGCTCTCTCCGCAGCAGCTGCCTCC
Phenotype
MIM: 601570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT7A PubMed Links

Gene Details

Gene
WNT7A
Gene Name
Wnt family member 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004625.3 Intron NP_004616.2
XM_011534091.2 Intron XP_011532393.1

View Full Product Details