Product Details

SNP ID
rs13326925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13823547 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAGGTGAAGTTGAGAGGATTCAA[A/G]GAAAAGCCATGGCAAAGGTGGGGGA
Phenotype
MIM: 601570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT7A PubMed Links

Gene Details

Gene
WNT7A
Gene Name
Wnt family member 7A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004625.3 Intron NP_004616.2
XM_011534091.2 Intron XP_011532393.1

View Full Product Details