Product Details

SNP ID
rs28698226
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:50432576 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
CAAACTGATAACAAACAGTTCCCCA[A/G]TCAGCACTGGTCATTGGACCATACT
Phenotype
MIM: 603158
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
USP8 PubMed Links

Gene Details

Gene
USP8
Gene Name
ubiquitin specific peptidase 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128610.2 Intron NP_001122082.1
NM_001283049.1 Intron NP_001269978.1
NM_005154.4 Intron NP_005145.3
XM_006720761.3 Intron XP_006720824.1
XM_006720762.3 Intron XP_006720825.1
XM_011522193.2 Intron XP_011520495.1
XM_017022718.1 Intron XP_016878207.1
XM_017022719.1 Intron XP_016878208.1
XM_017022720.1 Intron XP_016878209.1
XM_017022721.1 Intron XP_016878210.1
XM_017022722.1 Intron XP_016878211.1

View Full Product Details