Product Details

SNP ID
rs1017522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8222791 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTGCAGCGCTGGTCAGGTAAC[A/C]TATTCCTTGAAGACACTCTTGTCTC
Phenotype
MIM: 613129
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CTC1 PubMed Links

Gene Details

Gene
CTC1
Gene Name
CTS telomere maintenance complex component 1
There are no transcripts associated with this gene.

Gene
LINC00324
Gene Name
long intergenic non-protein coding RNA 324
There are no transcripts associated with this gene.

View Full Product Details