Product Details

SNP ID
rs1133221
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.17:47899023 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGATAAATGGACATCATTTTGGAA[G/T]GCATTTGTTGCTTTTTTGCTTGTTT
Phenotype
MIM: 601801
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC102724532 PubMed Links

Gene Details

Gene
LOC102724532
Gene Name
uncharacterized LOC102724532
There are no transcripts associated with this gene.

Gene
SP2
Gene Name
Sp2 transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003110.5 Intron NP_003101.3
XM_006722023.2 Intron XP_006722086.1
XM_006722025.2 Intron XP_006722088.1
XM_006722026.2 Intron XP_006722089.1
XM_006722027.2 Intron XP_006722090.1
XM_011525136.2 Intron XP_011523438.1
XM_011525137.2 Intron XP_011523439.1
XM_011525138.2 Intron XP_011523440.1
XM_011525139.2 Intron XP_011523441.1
XM_011525140.2 Intron XP_011523442.1
XM_011525142.1 Intron XP_011523444.1
XM_011525143.1 Intron XP_011523445.1
XM_017024968.1 Intron XP_016880457.1
XM_017024969.1 Intron XP_016880458.1
Gene
SP2-AS1
Gene Name
SP2 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details