Product Details

SNP ID
rs1438116
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.4:165334405 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TTTAAGCTAGCTGGAAAGGAAAGGT[A/G]TGGTGATTTGCGCATATTTTGCAGT
Phenotype
MIM: 607545
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MSMO1 PubMed Links

Gene Details

Gene
MSMO1
Gene Name
methylsterol monooxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017369.2 Intron NP_001017369.1
NM_006745.4 Intron NP_006736.1
XM_005263176.2 Intron XP_005263233.1

View Full Product Details