Product Details

SNP ID
rs1787725
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62814983 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGATATCACATATTAGGAAATA[G/A]TTAATTCTAACAGATAGGATAACTG
Phenotype
MIM: 602647 MIM: 603189
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
NXF1 PubMed Links

Gene Details

Gene
NXF1
Gene Name
nuclear RNA export factor 1
There are no transcripts associated with this gene.

Gene
STX5
Gene Name
syntaxin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244666.1 Intron NP_001231595.1
NM_003164.4 Intron NP_003155.2
XM_011545223.2 Intron XP_011543525.1
XM_011545224.2 Intron XP_011543526.1

View Full Product Details