Product Details
- SNP ID
-
rs2332240
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:122577987 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CTCTCTTCTAGGGGGCGCCGACTAC[A/G]CTGACTTCCCTGTTCCGGAAGAGGG
- Phenotype
-
MIM: 613143
MIM: 612066
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DTX3L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs141265268,rs190522327] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DTX3L
- Gene Name
- deltex E3 ubiquitin ligase 3L
There are no transcripts associated with this gene.
- Gene
- PARP15
- Gene Name
- poly(ADP-ribose) polymerase family member 15
View Full Product Details