Product Details

SNP ID
rs886177
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:109713885 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGACTGGAGACGTCTATAACCTG[A/G]TTCTCCAAAATGTCCACACGAATCT
Phenotype
MIM: 138385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSTM5 PubMed Links

Gene Details

Gene
GSTM5
Gene Name
glutathione S-transferase mu 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000851.3 Intron NP_000842.2
XM_005270784.4 Intron XP_005270841.1
XM_005270785.4 Intron XP_005270842.1

View Full Product Details