Product Details

SNP ID
rs1126673
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.4:99124465 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
CTTCAAAAGATGAGGATTGTTCGGA[C/T]GCTGTTAATAACAATAAAACATTAA
Phenotype
MIM: 103740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADH4 PubMed Links

Gene Details

Gene
ADH4
Gene Name
alcohol dehydrogenase 4 (class II), pi polypeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000670.4 1336 Missense Mutation ATC,GTC I374V NP_000661.2
NM_001306171.1 1336 Missense Mutation ATC,GTC I393V NP_001293100.1
NM_001306172.1 1336 Missense Mutation ATC,GTC I393V NP_001293101.1
XM_017007713.1 1336 Intron XP_016863202.1
XM_017007714.1 1336 Intron XP_016863203.1
XM_017007715.1 1336 Intron XP_016863204.1
Gene
LOC100507053
Gene Name
uncharacterized LOC100507053
There are no transcripts associated with this gene.

View Full Product Details