Product Details
- SNP ID
-
rs17552409
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:100398175 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCTAGGAACGCTCCTGTAAATGGA[A/G]TAATGGAATTTTTAAATGTGAGAGA
- Phenotype
-
MIM: 608350
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EMCN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74410782] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EMCN
- Gene Name
- endomucin
- Gene
- SNORA101A
- Gene Name
- small nucleolar RNA, H/ACA box 101A
There are no transcripts associated with this gene.
View Full Product Details