Product Details

SNP ID
rs1237554
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:192141196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAAAAGTGACGAATGAATTGAA[A/G]ACCCAGAAATCAGAACAATGAAAAT
Phenotype
MIM: 601513
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF12 PubMed Links

Gene Details

Gene
FGF12
Gene Name
fibroblast growth factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004113.5 3456 UTR 3 NP_004104.3
NM_021032.4 3456 UTR 3 NP_066360.1
XM_005247227.2 3456 UTR 3 XP_005247284.1
XM_006713538.3 3456 UTR 3 XP_006713601.1
XM_017005879.1 3456 UTR 3 XP_016861368.1

View Full Product Details