Product Details
- SNP ID
-
rs1516792
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:112778356 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAAATACTTGGTTAATGTTCTTA[A/G]AATTTTCTACGTTGAATGAATCCTT
- Phenotype
-
MIM: 147760
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IL1A
PubMed Links
Gene Details
- Gene
- IL1A
- Gene Name
- interleukin 1 alpha
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000575.4 |
|
Intron |
|
|
NP_000566.3 |
View Full Product Details