Product Details

SNP ID
rs1065435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:62909934 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGGGAACACAGTCCTTGCAGAA[C/G]GTGGACACCAGCACCATGATGTTGT
Phenotype
MIM: 118510
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CHRM1 PubMed Links

Gene Details

Gene
CHRM1
Gene Name
cholinergic receptor muscarinic 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000738.2 1269 Silent Mutation ACC,ACG T389T NP_000729.2
XM_011544742.2 1269 Silent Mutation ACC,ACG T389T XP_011543044.1

View Full Product Details