Product Details

SNP ID
rs1051120
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:67351639 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCTCTCAGATTCCTGGCCTTAGG[G/T]GGGAGAGCATCTTTTCCTCATGGAT
Phenotype
MIM: 611525
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC100130987 PubMed Links

Gene Details

Gene
LOC100130987
Gene Name
uncharacterized LOC100130987
There are no transcripts associated with this gene.

Gene
POLD4
Gene Name
DNA polymerase delta 4, accessory subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256870.1 765 UTR 3 NP_001243799.1
NM_021173.4 765 UTR 3 NP_066996.3

View Full Product Details