Product Details

SNP ID
rs11784963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:74828152 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGCCAGGTAAATAAATAAGTTACA[C/T]AGGATGTAAGATGATAAGGTTATGA
Phenotype
MIM: 607076
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PI15 PubMed Links

Gene Details

Gene
PI15
Gene Name
peptidase inhibitor 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001324403.1 Intron NP_001311332.1
NM_015886.4 Intron NP_056970.1

View Full Product Details