Product Details

SNP ID
rs1681073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:55902142 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGGGCTGTCTCTTCCAGGGACAC[C/T]TTATCAAGAGTCCTGCTCAAGGCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PYM1 PubMed Links

Gene Details

Gene
PYM1
Gene Name
PYM homolog 1, exon junction complex associated factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143853.1 497 Silent Mutation AAA,AAG K114K NP_001137325.1
NM_032345.2 497 Silent Mutation AAA,AAG K115K NP_115721.1

View Full Product Details