Product Details

SNP ID
rs1171138
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:22934322 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTCATTTGTCATAACATTAATAT[A/T]TGCAGATATAGTACTGATTACATTT
Phenotype
MIM: 611226
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ARMC3 PubMed Links

Gene Details

Gene
ARMC3
Gene Name
armadillo repeat containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282745.1 Intron NP_001269674.1
NM_001282746.1 Intron NP_001269675.1
NM_001282747.1 Intron NP_001269676.1
NM_173081.4 Intron NP_775104.2
XM_005252380.2 Intron XP_005252437.1
XM_005252381.2 Intron XP_005252438.1
XM_011519350.2 Intron XP_011517652.1
XM_011519351.2 Intron XP_011517653.1
XM_017015831.1 Intron XP_016871320.1
XM_017015832.1 Intron XP_016871321.1
XM_017015833.1 Intron XP_016871322.1

View Full Product Details