Product Details

SNP ID
rs1128396
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6614875 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCTTCCATACTTACATCAAAGAG[A/T]CCTGCCCCATGCTGCTGGTAGAGCC
Phenotype
MIM: 603057 MIM: 602366 MIM: 600475 MIM: 607998
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DCHS1 PubMed Links

Gene Details

Gene
DCHS1
Gene Name
dachsous cadherin-related 1
There are no transcripts associated with this gene.

Gene
ILK
Gene Name
integrin linked kinase
There are no transcripts associated with this gene.

Gene
TAF10
Gene Name
TATA-box binding protein associated factor 10
There are no transcripts associated with this gene.

Gene
TPP1
Gene Name
tripeptidyl peptidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000391.3 1603 Silent Mutation GGA,GGT G514G NP_000382.3

View Full Product Details