Product Details

SNP ID
rs1042043
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6259940 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCGGCGGGGGCCATGGAGCTGCT[A/C]AAGCTGAACCGGAGCGTGCAGGGAA
Phenotype
MIM: 118445
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CCKBR PubMed Links

Gene Details

Gene
CCKBR
Gene Name
cholecystokinin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318029.1 330 Silent Mutation CTA,CTC L4L NP_001304958.1
NM_176875.3 330 Silent Mutation CTA,CTC L4L NP_795344.1
XM_005253210.1 330 Silent Mutation CTA,CTC L4L XP_005253267.1
XM_017018516.1 330 Intron XP_016874005.1

View Full Product Details