Product Details

SNP ID
rs999074
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.12:7711716 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGGGTTGGGAGGTCAAAAGGCTG[C/T]CGTCTTTTTTTTTTTTTTTTTTTTT
Phenotype
MIM: 608408
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DPPA3 PubMed Links

Gene Details

Gene
DPPA3
Gene Name
developmental pluripotency associated 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_199286.3 Intron NP_954980.1

View Full Product Details