Product Details

SNP ID
rs1190761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56194154 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATCTGCTTGACATGTTCTTATTTTT[A/G]TTCGAGAGCTCCCTGGGGCTTTCTT
Phenotype
MIM: 139311
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DKFZP434H168 PubMed Links

Gene Details

Gene
DKFZP434H168
Gene Name
uncharacterized LOC26077
There are no transcripts associated with this gene.

Gene
GNAO1
Gene Name
G protein subunit alpha o1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020988.2 Intron NP_066268.1
NM_138736.2 Intron NP_620073.2
XM_011523003.2 Intron XP_011521305.1
Gene
LOC283856
Gene Name
uncharacterized LOC283856
There are no transcripts associated with this gene.

View Full Product Details