Product Details

SNP ID
rs1803104
Assay Type
Functionally Tested
NCBI dbSNP Submissions
1
Location
Chr.1:230702825 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTTTCACAAACAAGCTGGTCGG[C/T]TGGAATTCTTTTTGGAACAGTAGTC
Phenotype
MIM: 106150 MIM: 606974
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
AGT PubMed Links

Gene Details

Gene
AGT
Gene Name
angiotensinogen
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000029.3 2282 UTR 3 NP_000020.1
Gene
COG2
Gene Name
component of oligomeric golgi complex 2
There are no transcripts associated with this gene.

View Full Product Details