Product Details

SNP ID
rs1148454
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:13584898 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCTTCCATTACCCTTTGTCCTCTC[C/T]TGGAGCCTTACCTCTTCCCGCTGGG
Phenotype
MIM: 608863
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDPN PubMed Links

Gene Details

Gene
PDPN
Gene Name
podoplanin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001006624.1 Intron NP_001006625.1
NM_001006625.1 Intron NP_001006626.1
NM_006474.4 Intron NP_006465.3
NM_198389.2 Intron NP_938203.2
XM_006710295.1 Intron XP_006710358.1

View Full Product Details