Product Details

SNP ID
rs13409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:31164363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAGTGTGTCTATCTACTGTGTCCCA[A/G]GCTTCTTTATTTAAGAAAAAAGTGA
Phenotype
MIM: 605310 MIM: 164177 MIM: 600912
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCHCR1 PubMed Links

Gene Details

Gene
CCHCR1
Gene Name
coiled-coil alpha-helical rod protein 1
There are no transcripts associated with this gene.

Gene
POU5F1
Gene Name
POU class 5 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001173531.2 2039 UTR 3 NP_001167002.1
NM_001285986.1 2039 UTR 3 NP_001272915.1
NM_001285987.1 2039 UTR 3 NP_001272916.1
NM_002701.5 2039 Intron NP_002692.2
NM_203289.5 2039 UTR 3 NP_976034.4
Gene
PSORS1C3
Gene Name
psoriasis susceptibility 1 candidate 3 (non-protein coding)
There are no transcripts associated with this gene.

Gene
TCF19
Gene Name
transcription factor 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001077511.1 2039 Intron NP_001070979.1
NM_001318908.1 2039 Intron NP_001305837.1
NM_007109.2 2039 Intron NP_009040.2

View Full Product Details