Product Details
- SNP ID
-
rs1051431
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:123161256 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGACTGAAACCGGCTTACACTGTTC[A/G]TAGCTAACTGATTTATTCTTCTCCC
- Phenotype
-
MIM: 605501
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MPHOSPH9
PubMed Links
Gene Details
- Gene
- MPHOSPH9
- Gene Name
- M-phase phosphoprotein 9
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022782.3 |
3520 |
Silent Mutation |
TAC,TAT |
Y1087Y |
NP_073619.3 |
XM_011537740.2 |
3520 |
Silent Mutation |
TAC,TAT |
Y1087Y |
XP_011536042.1 |
XM_011537741.2 |
3520 |
Silent Mutation |
TAC,TAT |
Y1087Y |
XP_011536043.1 |
XM_017018673.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y1087Y |
XP_016874162.1 |
XM_017018674.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y1053Y |
XP_016874163.1 |
XM_017018675.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y1045Y |
XP_016874164.1 |
XM_017018676.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y1035Y |
XP_016874165.1 |
XM_017018677.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y1012Y |
XP_016874166.1 |
XM_017018678.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y978Y |
XP_016874167.1 |
XM_017018679.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y935Y |
XP_016874168.1 |
XM_017018680.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y935Y |
XP_016874169.1 |
XM_017018681.1 |
3520 |
Silent Mutation |
TAC,TAT |
Y714Y |
XP_016874170.1 |
View Full Product Details