Product Details

SNP ID
rs1256930
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:201468349 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTCCCCCGGGGGCTTACCTGCCTTG[A/T]CCTCAGCACTGAGGTGGTGGGTAGC
Phenotype
MIM: 607054
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PHLDA3 PubMed Links

Gene Details

Gene
PHLDA3
Gene Name
pleckstrin homology like domain family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012396.4 823 UTR 3 NP_036528.1

View Full Product Details