Product Details

SNP ID
rs1053592
Assay Type
Validated
NCBI dbSNP Submissions
67
Location
Chr.1:201465612 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GAACACCGCAGTGGGGGGAAGATGC[A/G]TAGAGGTGGGGGAAGAAGTGTGCAA
Phenotype
MIM: 607054
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHLDA3 PubMed Links

Gene Details

Gene
PHLDA3
Gene Name
pleckstrin homology like domain family A member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012396.4 1398 UTR 3 NP_036528.1

View Full Product Details