Product Details

SNP ID
rs17463552
Assay Type
Functionally tested
NCBI dbSNP Submissions
37
Location
Chr.1:182412700 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACACAGTGAGGCATTTAGTGGTAC[A/G]GACTCTAGGACTACTCCTGCCATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LINC00272 PubMed Links

Gene Details

Gene
LINC00272
Gene Name
long intergenic non-protein coding RNA 272
There are no transcripts associated with this gene.

View Full Product Details