Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001127204.1 | 12 | Intron | NP_001120676.1 | ||
NM_001127205.1 | 12 | Intron | NP_001120677.1 | ||
NM_001127206.2 | 12 | Intron | NP_001120678.1 | ||
NM_001286267.1 | 12 | Intron | NP_001273196.1 | ||
NM_001286268.1 | 12 | Intron | NP_001273197.1 | ||
NM_001286269.1 | 12 | Intron | NP_001273198.1 | ||
NM_001286270.1 | 12 | Intron | NP_001273199.1 | ||
NM_001286271.1 | 12 | Intron | NP_001273200.1 | ||
NM_002134.3 | 12 | Intron | NP_002125.3 | ||
XM_011522473.1 | 12 | Intron | XP_011520775.1 | ||
XM_017023196.1 | 12 | Intron | XP_016878685.1 | ||
XM_017023197.1 | 12 | Intron | XP_016878686.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001305141.1 | 12 | Intron | NP_001292070.1 | ||
NM_001305142.1 | 12 | Intron | NP_001292071.1 | ||
NM_020677.4 | 12 | Intron | NP_065728.1 | ||
XM_006720905.3 | 12 | UTR 5 | XP_006720968.1 | ||
XM_006720906.2 | 12 | Intron | XP_006720969.1 | ||
XM_011522567.2 | 12 | Intron | XP_011520869.1 | ||
XM_017023485.1 | 12 | Intron | XP_016878974.1 | ||
XM_017023486.1 | 12 | Intron | XP_016878975.1 | ||
XM_017023487.1 | 12 | Intron | XP_016878976.1 | ||
XM_017023488.1 | 12 | Intron | XP_016878977.1 | ||
XM_017023489.1 | 12 | Intron | XP_016878978.1 | ||
XM_017023490.1 | 12 | Intron | XP_016878979.1 | ||
XM_017023491.1 | 12 | Intron | XP_016878980.1 | ||
XM_017023492.1 | 12 | Intron | XP_016878981.1 |