Product Details

SNP ID
rs1773218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:194686291 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGGAGGTGGGGAGCACCGGCCCCT[A/G]GGCACACTCGCTGTAGAGTTTCCGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM43A PubMed Links

Gene Details

Gene
FAM43A
Gene Name
family with sequence similarity 43 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153690.4 399 UTR 5 NP_710157.2

View Full Product Details