Product Details

SNP ID
rs3752798
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:194686676 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TCCTAAGCACTCTCTCTGCTCCCCT[A/C]CCCCCAACTCCCTACCACAGGGCCG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM43A PubMed Links

Gene Details

Gene
FAM43A
Gene Name
family with sequence similarity 43 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153690.4 784 UTR 5 NP_710157.2

View Full Product Details