Product Details

SNP ID
rs7522439
Assay Type
Functionally tested
NCBI dbSNP Submissions
29
Location
Chr.1:17250899 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGGATGGATTGGGGCAATAAGAG[A/G]TAAAGGCCATAGCTTCCTGGAGTCA
Phenotype
MIM: 607934 MIM: 606755
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PADI1 PubMed Links

Gene Details

Gene
PADI1
Gene Name
peptidyl arginine deiminase 1
There are no transcripts associated with this gene.

Gene
PADI3
Gene Name
peptidyl arginine deiminase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016233.2 Intron NP_057317.2
XM_011541571.2 Intron XP_011539873.1
XM_011541572.2 Intron XP_011539874.1
XM_017001463.1 Intron XP_016856952.1

View Full Product Details