Product Details

SNP ID
rs6932865
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.6:26539938 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GTTATTATGGTGCCCAGAAAGGGGG[A/C]AAAGGTTCCAGGGTCCTAGGAGATG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HMGN4 PubMed Links

Gene Details

Gene
HMGN4
Gene Name
high mobility group nucleosomal binding domain 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006353.2 193 Intron NP_006344.1
XM_017010171.1 193 Intron XP_016865660.1
XM_017010172.1 193 UTR 5 XP_016865661.1
XM_017010173.1 193 Intron XP_016865662.1

View Full Product Details