Product Details

SNP ID
rs3813922
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:32189533 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTATCACCTTGTTCTGGAAGTAG[G/T]GGTTGCTCCAAAAGCGGAACTTGAA
Phenotype
MIM: 604866
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PLAGL2 PubMed Links

Gene Details

Gene
PLAGL2
Gene Name
PLAG1 like zinc finger 2
There are no transcripts associated with this gene.

Gene
TSPY26P
Gene Name
testis specific protein, Y-linked 26, pseudogene
There are no transcripts associated with this gene.

View Full Product Details