Product Details
- SNP ID
-
rs7840284
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:94723750 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- TTGATATGTCTTAAAATCAGTGCAC[A/G]CATTCAGTATGACAGTGTGGTTTTT
- Phenotype
-
MIM: 613895
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DPY19L4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76627971] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DPY19L4
- Gene Name
- dpy-19 like 4 (C. elegans)
View Full Product Details