Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199397.1 | 4852 | UTR 3 | NP_001186326.1 | ||
NM_001199398.1 | 4852 | UTR 3 | NP_001186327.1 | ||
NM_001199399.1 | 4852 | UTR 3 | NP_001186328.1 | ||
NM_001199400.1 | 4852 | UTR 3 | NP_001186329.1 | ||
NM_012224.2 | 4852 | UTR 3 | NP_036356.1 | ||
XM_006714228.1 | 4852 | UTR 3 | XP_006714291.1 | ||
XM_011532003.1 | 4852 | UTR 3 | XP_011530305.1 | ||
XM_011532004.1 | 4852 | UTR 3 | XP_011530306.1 | ||
XM_011532005.2 | 4852 | Intron | XP_011530307.1 | ||
XM_017008249.1 | 4852 | UTR 3 | XP_016863738.1 | ||
XM_017008250.1 | 4852 | UTR 3 | XP_016863739.1 | ||
XM_017008251.1 | 4852 | UTR 3 | XP_016863740.1 | ||
XM_017008252.1 | 4852 | UTR 3 | XP_016863741.1 | ||
XM_017008253.1 | 4852 | UTR 3 | XP_016863742.1 | ||
XM_017008254.1 | 4852 | UTR 3 | XP_016863743.1 |