Product Details

SNP ID
rs35607186
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:55994052 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAACTTATGCTTTCTTTCAGGATTG[C/T]GTCAGAACAAGAGAGCTTGAAAAGT
Phenotype
MIM: 608492
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR5F1 PubMed Links

Gene Details

Gene
OR5F1
Gene Name
olfactory receptor family 5 subfamily F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003697.1 574 Missense Mutation ACA,GCA T192A NP_003688.1
Gene
OR7E5P
Gene Name
olfactory receptor family 7 subfamily E member 5 pseudogene
There are no transcripts associated with this gene.

View Full Product Details