Product Details

SNP ID
rs6061243
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:62465397 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGACGCCAGGCTGGGCTTGGCTTT[C/G]GAAGTGGCTGCTGAGCTGTCAGTGC
Phenotype
MIM: 611496
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GATA5 PubMed Links

Gene Details

Gene
GATA5
Gene Name
GATA binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080473.4 1043 Silent Mutation TCC,TCG S327S NP_536721.1
XM_006723699.3 1043 Silent Mutation TCC,TCG S327S XP_006723762.1

View Full Product Details